Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040535.2 | NP_001035625.1 | 2 - 3 | 180 | 74 | |
GenBank mRNA | BC109989.1 | - | 2 - 3 | 182 | 74 | |
DQ677839.1 | - | 1 - 2 | 231 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |