Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040551.2 | NP_001035641.1 | 5 - 6 | 708 | 148 | |
GenBank mRNA | BC109499.1 | - | 5 - 6 | 709 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |