Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040571.2 | NP_001035661.1 | 5 - 6 | 572 | 99 | |
GenBank mRNA | BC109581.1 | - | 5 - 6 | 573 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |