Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045932.1 | NP_001039397.1 | 4 - 5 | 418 | 74 | |
GenBank mRNA | BC110165.1 | - | 4 - 5 | 418 | 74 | |
BC149132.1 | - | 3 - 4 | 772 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |