Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045933.1 | NP_001039398.1 | 9 - 10 | 1048 | 119 | |
GenBank mRNA | BC149282.1 | - | 12 - 13 | 1456 | 119 | |
DQ322170.1 | - | 9 - 10 | 1048 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |