Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045945.1 | NP_001039410.1 | 3 - 4 | 516 | 64 | |
GenBank mRNA | BC112714.1 | - | 3 - 4 | 516 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |