Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045953.2 | NP_001039418.2 | 5 - 6 | 760 | 106 | |
GenBank mRNA | BC112670.1 | - | 5 - 6 | 716 | 106 | |
EV657492.1 | - | 5 - 6 | 767 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |