Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046081.2 | NP_001039546.1 | 2 - 3 | 335 | 101 | |
GenBank mRNA | BC110157.1 | - | 2 - 3 | 336 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |