Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECSCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046099.2 | NP_001039564.1 | 4 - 5 | 406 | 71 | |
NM_001244445.1 | NP_001231374.1 | 6 - 7 | 511 | 71 | ||
NM_001244446.1 | NP_001231375.1 | 5 - 6 | 460 | 71 | ||
GenBank mRNA | BC112471.1 | - | 4 - 5 | 395 | 71 | |
DV851687.1 | - | 6 - 7 | 509 | 71 | ||
EV675062.1 | - | 5 - 6 | 463 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |