Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046104.2 | NP_001039569.1 | 6 - 7 | 723 | 138 | |
GenBank mRNA | BC112535.1 | - | 6 - 7 | 724 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |