Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046188.2 | NP_001039653.1 | 11 - 12 | 1264 | 150 | |
GenBank mRNA | BC111184.1 | - | 11 - 12 | 1265 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |