Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046285.1 | NP_001039750.1 | 1 - 2 | 146 | 157 | |
GenBank mRNA | BC112635.1 | - | 1 - 2 | 146 | 157 | |
BT025423.1 | - | 1 - 2 | 174 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |