Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046306.2 | NP_001039771.1 | 5 - 6 | 855 | 75 | |
GenBank mRNA | BC112835.1 | - | 5 - 6 | 856 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |