Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046334.1 | NP_001039799.1 | 5 - 6 | 743 | 105 | |
GenBank mRNA | BC105411.1 | - | 5 - 6 | 743 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |