Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046390.2 | NP_001039855.1 | 2 - 3 | 278 | 109 | |
GenBank mRNA | BC110254.1 | - | 2 - 3 | 279 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |