Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046390.2 | NP_001039855.1 | 3 - 4 | 532 | 57 | |
GenBank mRNA | BC110254.1 | - | 3 - 4 | 533 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |