Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046390.2 | NP_001039855.1 | 5 - 6 | 1082 | 97 | |
GenBank mRNA | BC110254.1 | - | 5 - 6 | 1083 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |