Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046454.1 | NP_001039919.1 | 5 - 6 | 493 | 54 | |
GenBank mRNA | BC110170.1 | - | 5 - 6 | 493 | 54 | |
HE864424.1 | - | 4 - 5 | 378 | 54 | ||
HE864487.1 | - | 4 - 5 | 378 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |