Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046502.2 | NP_001039967.1 | 3 - 4 | 829 | 94 | |
GenBank mRNA | BC105377.1 | - | 3 - 4 | 831 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |