Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046542.2 | NP_001040007.1 | 11 - 12 | 1231 | 117 | |
GenBank mRNA | BC112463.1 | - | 11 - 12 | 1232 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |