Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046542.2 | NP_001040007.1 | 9 - 10 | 985 | 80 | |
GenBank mRNA | BC112463.1 | - | 9 - 10 | 986 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |