Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075141.2 | NP_001068609.2 | 8 - 9 | 734 | 134 | |
NM_001257088.1 | NP_001244017.1 | 7 - 8 | 616 | 134 | ||
GenBank mRNA | BC105495.1 | - | 7 - 8 | 2069 | 134 | |
FE012554.1 | - | 7 - 8 | 523 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |