Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075539.2 | NP_001069007.1 | 6 - 7 | 531 | 131 | |
GenBank mRNA | BC109525.1 | - | 6 - 7 | 532 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |