Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045536.1 | 1 - 2 | 30 | 73 | ||
GenBank mRNA | BC110259.1 | - | 1 - 2 | 31 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |