Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075604.2 | NP_001069072.2 | 3 - 4 | 252 | 84 | |
NR_045536.1 | 3 - 4 | 393 | 84 | |||
GenBank mRNA | BC110259.1 | - | 3 - 4 | 394 | 84 | |
DT858178.1 | - | 3 - 4 | 253 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |