Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075609.2 | NP_001069077.1 | 7 - 8 | 543 | 109 | |
GenBank mRNA | BC109751.1 | - | 7 - 8 | 544 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |