Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075763.2 | NP_001069231.1 | 11 - 12 | 1738 | 106 | |
GenBank mRNA | BC111238.1 | - | 11 - 12 | 1740 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |