Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075774.2 | NP_001069242.1 | 3 - 4 | 296 | 161 | |
GenBank mRNA | BC109646.1 | - | 3 - 4 | 297 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |