Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075774.2 | NP_001069242.1 | 4 - 5 | 411 | 90 | |
GenBank mRNA | BC109646.1 | - | 4 - 5 | 412 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |