Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075774.2 | NP_001069242.1 | 6 - 7 | 568 | 66 | |
GenBank mRNA | BC109646.1 | - | 6 - 7 | 569 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |