Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075774.2 | NP_001069242.1 | 8 - 9 | 821 | 84 | |
GenBank mRNA | BC109646.1 | - | 8 - 9 | 822 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |