Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075797.3 | NP_001069265.2 | 6 - 7 | 738 | 145 | |
GenBank mRNA | BC109850.1 | - | 6 - 7 | 737 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |