Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075797.3 | NP_001069265.2 | 7 - 8 | 851 | 78 | |
GenBank mRNA | BC109850.1 | - | 7 - 8 | 850 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |