Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075916.2 | NP_001069384.1 | 1 | 949 | 90 | |
GenBank mRNA | BC109745.1 | - | 1 | 950 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |