Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076240.2 | NP_001069708.1 | 2 - 3 | 447 | 68 | |
GenBank mRNA | BC109997.1 | - | 2 - 3 | 447 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |