Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076245.2 | NP_001069713.1 | 6 - 7 | 882 | 99 | |
GenBank mRNA | BC109679.1 | - | 6 - 7 | 885 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |