Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076378.2 | NP_001069846.1 | 6 - 7 | 789 | 80 | |
GenBank mRNA | BC109728.1 | - | 6 - 7 | 790 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |