Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076438.2 | NP_001069906.1 | 7 - 8 | 724 | 83 | |
GenBank mRNA | BC109690.1 | - | 7 - 8 | 726 | 83 | |
BC142401.1 | - | 7 - 8 | 583 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |