Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H7orf61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076486.2 | NP_001069954.1 | 2 - 3 | 605 | 70 | |
GenBank mRNA | BC111222.1 | - | 2 - 3 | 606 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |