Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076956.1 | NP_001070424.1 | 1 | 2444 | 119 | |
GenBank mRNA | BC111192.1 | - | 1 | 2444 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |