Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077047.2 | NP_001070515.1 | 4 - 5 | 574 | 147 | |
GenBank mRNA | BC105491.1 | - | 4 - 5 | 575 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |