Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077053.2 | NP_001070521.1 | 4 - 5 | 356 | 89 | |
GenBank mRNA | BC109709.1 | - | 4 - 5 | 357 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |