Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077058.2 | NP_001070526.1 | 2 - 3 | 184 | 76 | |
GenBank mRNA | BC109842.1 | - | 2 - 3 | 185 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |