Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H19orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077082.2 | NP_001070550.1 | 3 - 4 | 452 | 97 | |
GenBank mRNA | BC111650.1 | - | 3 - 4 | 453 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |