Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077977.2 | NP_001071445.1 | 1 - 2 | 73 | 85 | |
GenBank mRNA | BC112873.1 | - | 1 - 2 | 74 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |