Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078038.1 | NP_001071506.1 | 6 - 7 | 771 | 114 | |
GenBank mRNA | BC111261.1 | - | 6 - 7 | 771 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |