Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H16orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078045.1 | NP_001071513.1 | 1 - 2 | 193 | 147 | |
GenBank mRNA | BC109943.1 | - | 1 - 2 | 193 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |