Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079783.1 | NP_001073251.1 | 5 - 6 | 612 | 123 | |
GenBank mRNA | BC111218.1 | - | 5 - 6 | 612 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |