Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079799.1 | NP_001073267.1 | 7 - 8 | 891 | 56 | |
GenBank mRNA | BC112534.1 | - | 7 - 8 | 891 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |