Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174253.3 | NP_776678.2 | 3 - 4 | 610 | 129 | |
GenBank mRNA | BC112543.1 | - | 3 - 4 | 613 | 129 | |
Z31694.1 | - | 494 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |